HYGEN™ Quick Connect Single-Sided Frame, Rubbermaid® Commercial
Supplier: Janitorial Supplies
Certificates
About this item
The Rubbermaid Commercial Products HYGEN Mop Quick-Connect Wet/Dry Single-Sided Frame features a flat profile that slides easily under furniture and equipment.
- For bare floors
- Connects to microfiber mop heads with a hook-and-loop fastener that can be replaced when worn
- Features a nylon swivel joint for flexible maneuvering
- Corrosion resistance
- Made with non-magnetic metals, enabling use near magnetically sensitive equipment, such as MRIs
The Rubbermaid HYGEN microfiber mop features a lightweight aluminum with stainless steel construction for improved productivity. The one-step connecting mechanism speeds the mop parts assembly and changeover. The flat frame profile is sanitary and easy to clean and the replaceable hook-and-loop strips are cost efficient and extend product life.
Frequently Bought Together