5X Detector™ Block, KPL Inc.
Catalog # 95059-480
Supplier: KPL
CAS Number:
Specifications
- Description:5X Detector™ Block Solution
- Size:240 mL
- Cat. No.:95059-480
- Supplier no.:71-83-00
Specifications
About this item
Detector Block is recommended as a general blocking agent for use in membrane-based detection assays
Detector Block can be used as both a blocking solution and a conjugate diluent in membrane applications such as Western, Northern, and Southern blotting. It is especially useful when traditional blocking solutions (i.e., milk, BSA, casein) reduce sensitivity or do not adequately block background.