Order Entry
United States
ContactUsLinkComponent
2-Cyclohexylethanol for synthesis, Sigma-Aldrich®
Irritant
2-Cyclohexylethanol for synthesis, Sigma-Aldrich®
Catalog # EM8.18556.0100
Supplier:  MilliporeSigma
CAS Number:  
Irritant
2-Cyclohexylethanol for synthesis, Sigma-Aldrich®
Catalog # EM8.18556.0100
Supplier:  MilliporeSigma
Supplier Number:  8.18556.0100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 mL
  • Cat. No.:
    EM8.18556.0100
  • Supplier no.:
    8.18556.0100

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -20 °C
  • Molar Mass:
    128.21 g/mol
  • Boiling Point:
    206 - 207 °C
  • Vapor Pressure:
    0.2 hPa (25 °C)
  • Flash Point:
    93 °C
  • Refractive Index:
    1.4641 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 - 6.3 %(V)
  • Density:
    0.92 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :128.21 g/mol
 :M-Clarity™ quality level = MQ200
 :GHS07
CAS Number:4442-79-9
Formula:C₈H₁₆O
MDL Number:MFCD00001525