
1-Bromooctane for synthesis, Sigma-Aldrich®
Catalog # EM8.01969.0250
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:250 ml
- Cat. No.:EM8.01969.0250
- Supplier no.:8.01969.0250
Specifications
Specification Test Results
- Solubility:0.002 g/l (20 °C)
- Melting Point:-55 °C
- Molar Mass:193.12 g/mol
- Boiling Point:201 - 202 °C (1013 mbar)
- Vapor Pressure:0.47 hPa (25 °C)
- Flash Point:80 °C
- Refractive Index:1.4503 (25 °C, 589 nm)
- Density:1.11 g/cm3 (20 °C)
Specification Test Results
About this item
:
:193.13 g/mol
Merck Index:12,06861
Boiling Pt:198…201 °C (1013 hPa)
Melting Pt:–55 °C
Density:1.112 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:203-912-9
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS09
CAS Number:111-83-1
Formula:BrH₂C(CH₂)₆CH₃
MDL Number:MFCD00000276