Order Entry
United States
ContactUsLinkComponent
 
Flammable
Health
Irritant
Butyl glycidyl ether for synthesis, Sigma-Aldrich®
Catalog # EM8.01553.0250
Supplier:  MilliporeSigma
CAS Number:  
Flammable
Health
Irritant
Butyl glycidyl ether for synthesis, Sigma-Aldrich®
Catalog # EM8.01553.0250
Supplier:  MilliporeSigma
Supplier Number:  8.01553.0250
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 ml
  • Cat. No.:
    EM8.01553.0250
  • Supplier no.:
    8.01553.0250

Specifications

Specification Test Results

  • Solubility:
    20 g/l (20 °C)
  • Molar Mass:
    130.19 g/mol
  • Boiling Point:
    163 - 165 °C (1013 mbar)
  • Vapor Pressure:
    5 hPa (20 °C)
  • Flash Point:
    43 °C
  • Density:
    0.92 g/cm3 (20 °C)
  • Ignition Point:
    215 °C

Specification Test Results

About this item

 :
 :130.19 g/mol
ADR:3,III
 :M-Clarity™ quality level = MQ200
EINECS:219-376-4
UN:3271
 :GHS02
 :GHS08
 :GHS07
CAS Number:2426-08-6
Formula:C₇H₁₄O₂
MDL Number:MFCD00005145