ContactUsLinkComponent
Copper Cell Test Method: photometric 0.05 - 8.00 mg/L Cu Spectroquant®
Copper Cell Test Method: photometric 0.05 - 8.00 mg/L Cu Spectroquant®
Catalog # EM1.14553.0001
Supplier:  MilliporeSigma
CAS Number:  
Copper Cell Test Method: photometric 0.05 - 8.00 mg/L Cu Spectroquant®
Catalog # EM1.14553.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14553.0001
CAS Number:  
Specifications
  • Chemical method:
    Cuprizone
  • Description:
    Copper cell test*7-3
  • Size:
    1 pc
  • Cat. No.:
    EM1.14553.0001
  • Range:
    0,05 - 8,00 mg/L Cu
  • Supplier no.:
    1.14553.0001
  • No. of tests:
    25
  • For unit:
    Prove, Nova 30/60, Multy, Move