
Zirconyl chloride octahydrate, EMSURE® for analysis, Supelco®
Catalog # EM1.08917.0100
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:EM1.08917.0100
- Supplier no.:1.08917.0100
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:322.28 g/mol
- Bulk Density:800 kg/m3
- Density:1.91 g/cm3 (20 °C)
- pH:1 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:322.25 g/mol
UN:3260
ADR:8,II
:GHS05
CAS Number:13520-92-8
Formula:Cl₂OZr·8H₂O
MDL Number:MFCD00149898