$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Environmental Test Kits
AOX Enrichment Set Spectroquant®
AOX Enrichment Set Spectroquant®
Catalog # EM1.00678.0001
Supplier:
MilliporeSigma
CAS Number:
SDS
Expand image
AOX Enrichment Set Spectroquant®
Catalog # EM1.00678.0001
Supplier:
MilliporeSigma
Supplier Number:
1.00678.0001
CAS Number:
SDS
Specifications
Pack type:
Plastic Bag(s)
Size:
1 pc
Cat. No.:
EM1.00678.0001
Supplier no.:
1.00678.0001
Specifications
About this item
Product Details
Documentation
Product Details
Documentation