C. difficile Toxoid B, Enzo Life Sciences
50 µg
Catalog # 10033-556
Supplier: Enzo Life Sciences
Supplier #: BML-G155-0050
<i>C. difficile</i> Toxoid B, Enzo Life Sciences
Catalog # 10033-556
Supplier: Enzo Life Sciences
CAS Number:
Jump to:
Product Details & Documents
From Clostridium difficile, strain VPI10463
Inactivated by formaldehyde.
Specifications
Specifications
- Size:50 µg
- Purity Detail:Purified protein.
- Cat. No.:10033-556
- Formulation:Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose.
- Purity:>95% (SDS-PAGE)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



