C. difficile Toxoid A, Enzo Life Sciences
Catalog # 10033-554
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:50 µg
- Purity Detail:Purified protein.
- Purity:>95% (SDS-PAGE)
- Formulation:Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose.
- Cat. No.:10033-554
- Supplier no.:BML-G145-0050
Specifications
About this item
From Clostridium difficile, strain VPI10463
Inactivated by formaldehyde.