Order Entry
United States
ContactUsLinkComponent
Square Plastic Sample Jug with Screw Cap, Wheaton, DWK Life Sciences
Square Plastic Sample Jug with Screw Cap, Wheaton, DWK Life Sciences
Catalog # 16300-256
Small Business Enterprise
undefined
Square Plastic Sample Jug with Screw Cap, Wheaton, DWK Life Sciences
Catalog # 16300-256
Supplier Number:  222333
Small Business Enterprise

Specifications

  • Cap Size:
    38-400
  • Capacity:
    4 L (1 gal.)
  • Cat. No.:
    16300-256
  • O.D.×H:
    153 x 285 mm
  • Cap color:
    White
  • Supplier no.:
    222333

Specifications

About this item

Wheaton square plastic sample jugs have good chemical resistance.

  • High density polyethylene

Narrow mouth design is ideal for storing and pouring liquids. Jugs include a pre-attached white polypropylene screw cap with PVC liner.