ContactUsLinkComponent
VWR® Heavy-Duty Equipment Clamps
  14216-208
Product available on GSA Advantage®
undefined
VWR® Heavy-Duty Equipment Clamps
  14216-208
Product available on GSA Advantage®
  • Description:
    Heavy Duty Clamps
  • Length:
    101.6 mm
  • Environmentally Preferable:
  • Cat. No.:
    14216-208
These heavy-duty clamps support overhead mixers and homogenizers.
  • Strong aluminum alloy construction
  • Long surface length
  • Easy-to-grip knobs
They fit rods from 6 to 24 mm in diameter. Large, easy-to-grip knobs allow extra tightening. Clamps are made of strong aluminum alloy with brushed finish. Long surface length supports rods to avoid vibration and wobble.