Order Entry
United States
ContactUsLinkComponent
SP Bel-Art Single Tube Holder Racks, Bel-Art Products, a part of SP
SP Bel-Art Single Tube Holder Racks, Bel-Art Products, a part of SP
Catalog # 10600-006
SP Bel-Art Single Tube Holder Racks, Bel-Art Products, a part of SP
Catalog # 10600-006
Supplier Number:  F18794-0003

Specifications

  • Description:
    Single Tube Holder
  • Dimensions:
    76 x 64 mm
  • Environmentally Preferable:
  • Cat. No.:
    10600-006
  • For tubes:
    50 ml
  • Supplier no.:
    187940003

Specifications

About this item

This holder solves the problem of where to put a single 50 mL conical tube where it will not roll or spill.

  • Epoxy-coated stainless steel construction
  • Chemical- and corrosion-resistant
  • Autoclavable

Steel wire with epoxy coating is chemical and corrosion resistant and steam autoclavable at 121°C.