ContactUsLinkComponent
Mini-Lab Flask Head, PTFE-Clad Bottom Joint, Ace Glass Incorporated
Mini-Lab Flask Head, PTFE-Clad Bottom Joint, Ace Glass Incorporated
Catalog # 89074-566
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Mini-Lab Flask Head, PTFE-Clad Bottom Joint, Ace Glass Incorporated
Catalog # 89074-566
Supplier:  Ace Glass
Supplier Number:  9446-10
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • [ST] joint no.:
    45/50 (inner)
  • Description:
    Flask Head
  • Cat. No.:
    89074-566
  • Center neck [ST] joint:
    19/38
  • Side neck ST joints:
    14/20
  • Supplier no.:
    9446-10
Specifications
About this item
Interchangeable Mini-Lab head with a standard taper 45/50 bottom joint
Top standard taper 19/38 center joint, and a standard taper 14/20 side necks. Side arms are placed at proper angles so that a thermometer or addition tube may be placed in the flask without interfering with the stirring. For use with cetain Ace flasks. PTFE-clad bottom joint.