Order Entry
United States
ContactUsLinkComponent
SP Bel-Art Magnetic Stirring Bars, PTFE, Bel-Art Products, a part of SP
SP Bel-Art Magnetic Stirring Bars, PTFE, Bel-Art Products, a part of SP
Catalog # 47744-998
SP Bel-Art Magnetic Stirring Bars, PTFE, Bel-Art Products, a part of SP
Catalog # 47744-998
Supplier Number:  37119-9003

Specifications

  • Coating:
    PTFE
  • Ø:
    10 mm (3/8")
  • Length:
    3 mm (⅛")
  • Cat. No.:
    47744-998
  • Supplier no.:
    371199003

Specifications

About this item

Stir bar with removable stabilizing ring to keep it centered in vessel.

  • Removable stabilizing ring
  • PTFE construction
  • Durable
  • Resists spinout

Creates deep vortex without splashing or chattering and resists spinout.