Instatherm® Large Oil Bath, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Large, borosilicate glass vessel coated with Ace Instatherm®
Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.
Can accept round-bottom flasks up to 5L, and other large vessels. The bath has a rolled lip for easy handling. The maximum temperature on this bath is 100°C. The full capacity is 15L without a vessel, and typically should have a minimum of six liters when using a vessel like a 5L round bottom flask. Baths must be run with a heating media or oil. Three-pronged plug for insertion into Ace temperature controllers.
Ordering information: Ace temperature controller is not included and sold separately; contact your Avantor representative for more information. Available as bath alone (with integral six-foot controller cord) or complete with clip for sensor and/or thermometer.
Frequently Bought Together