ContactUsLinkComponent
Micro/Mini-Lab Distillate Take-Off Adapter, Ace Glass Incorporated
  89075-146
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Micro/Mini-Lab Distillate Take-Off Adapter, Ace Glass Incorporated
  89075-146
 :  Ace Glass
 :  9563-07
Product available on GSA Advantage®
Small Business Enterprise
  • Cat. No.:
    89075-146
  • Top [ST] joint size:
    [ST] 14/10
  • Supplier no.:
    9563-07
The top joint of this adapter is a combination cap, thread and a standard taper joint to accommodate and seal other microscale apparatus.
  • O.D. of top joint has an external thread
  • I.D. at top is a ground [ST] outer joint
  • 105° angle
  • Cap and o-ring included
The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. 105° angle. Supplied with cap and o-ring.