Meisterhand Scapel Knife Handles, No. 3 and 3L, Integra™ Miltex®
Supplier: Miltex
Certificates
About this item
Fits surgical blades numbers 10-15c
Extra fine. Handel 3L is used for deep surgery.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.