Dimethyl sulfoxide, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Frequently Bought Together
KIMAX® Capillary Melting Point Tubes, Kimble Chase,DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
VWR® Disposable Transfer Pipettes, Graduated
Supplier: VWR International
Disposable Scalpels, Sterile, Integra™ Miltex®
Supplier: Miltex
Falcon® Tissue Culture Dishes, Polystyrene, Sterile, Corning
Supplier: Corning
Ethanol ≥70% (v/v), Biotechnology Grade
Supplier: VWR
Ethylene glycol ≥99.0% (by GC), AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Supplier: BeanTown Chemical
Blasticidin S hydrochloride solution 10 mg/ml in HEPES ≥95% (by HPLC)
Supplier: Invivogen
RQ1 RNase-Free DNase
Supplier: Promega Corporation
Anti-Rabbit Detection Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
LB Agar Plate with Ampicillin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Pierce™ Silver Stain Kit, Thermo Scientific
Supplier: Invitrogen
Corning® Fetal Bovine Serum, Regular, USDA Approved, Corning
Supplier: Corning
VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Supplier: VWR International
SepMate™-15 (IVD)
Supplier: STEMCELL Technologies
Logitech® M185 Wireless Mouse, Essendant LLC MS
Supplier: Janitorial Supplies