


Cobalt(II) chloride hexahydrate 98-102% ACS
Supplier: VWR
Certificates
About this item
CAS Number:7791-13-1
:
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652
:237.93 g/mol
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS05
Specification Test Results
- Assay:98% - 102%
- Calcium:= 0.005%
- Copper:= 0.002%
- Insolubles:= 0.01%
- Iron:= 0.005%
- Magnesium:= 0.005%
- Nickel:= 0.1%
- Nitrate:= 0.01%
- Potassium:= 0.01%
- Sodium:= 0.05%
- Sulfate:= 0.01%
- Zinc:= 0.03%
Specification Test Results
Related Information
Frequently Bought Together