Sodium hydroxide 0.01 N in aqueous solution
Plastic bottle, clear, 500 ml
Supplier: Aqua Solutions
: 8608-500ML
Sodium hydroxide 0.01 N in aqueous solution
Catalog # 101223-050
Supplier: Aqua Solutions
Marketsource
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1824
- ADR:8,III
Specifications
Specifications
- Pack type:Plastic bottle, clear
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



