Order Entry
United States
ContactUsLinkComponent
Amberlyst® 15 H⁺ form, Supelco®
Amberlyst® 15 H⁺ form, Supelco®
Catalog # 52428-696
Supplier:  MilliporeSigma
CAS Number:  
undefined
Amberlyst® 15 H⁺ form, Supelco®
Catalog # 52428-696
Supplier:  MilliporeSigma
Supplier Number:  1.15635.0500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 ml
  • Cat. No.:
    52428-696
  • Supplier no.:
    1.15635.0500

Specifications

Specification Test Results

  • Bulk Density:
    690 kg/m3
  • Density:
    0.6 g/cm3
  • pH:
    (H2O) not applicable

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:39389-20-3
MDL Number:MFCD00145841