Order Entry
United States
ContactUsLinkComponent
 
 
 
 
Nitric acid 69%, EMPARTA® analytical reagent, Supelco®
  EM1.01832.2500
 :  
 
 
 
Nitric acid 69%, EMPARTA® analytical reagent, Supelco®
  EM1.01832.2500
 :  1.01832.2500
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    EM1.01832.2500
  • Supplier no.:
    1.01832.2500

 

 

  • Appearance:
    passes test
  • As (Arsenic):
    ≤ 0.010 ppm
  • Assay (acidimetric):
    68.0 - 70.0 %
  • Chloride (Cl):
    ≤ 0.5 ppm
  • Colour:
    ≤ 10 Hazen
  • Fe (Iron):
    ≤ 0.200 ppm
  • Heavy metals (as Pb):
    ≤ 0.2 ppm
  • Identity:
    passes test
  • Residue on ignition (as SO₄):
    ≤ 5 ppm
  • Sulphate (SO₄):
    ≤ 1 ppm
  • Sulfate (SO₄):
    ≤ 1 ppm
  • Titratable base:
    ≤ 0.0004 meq/g

 

 

 :
 :63.01
 :13,06608
 :120.5 °C (1013 hPa)
 :–42 °C
 :1.4 g/cm³ (20 °C)
 :Ambient
 :231-714-2
 :2031
 :8,II
 :M-Clarity™ quality level = MQ200
 :GHS03
 :GHS06
 :GHS05
 :7697-37-2
 :HNO₃
 :MFCD00011349