ContactUsLinkComponent
SP Bel-Art Stopper/Tubing Sizer, Bel-Art Products, a part of SP
SP Bel-Art Stopper/Tubing Sizer, Bel-Art Products, a part of SP
Catalog # 59594-405
undefined
SP Bel-Art Stopper/Tubing Sizer, Bel-Art Products, a part of SP
Catalog # 59594-405
Supplier Number:  F13392-0000
Specifications
  • Description:
    Stopper sizer
  • L×W:
    15.6×6.6 cm (61/8 × 25/8")
  • Cat. No.:
    59594-405
  • Supplier no.:
    F133920000
Specifications
About this item
Handy stopper/tubing sizer easily determines the correct size of the cork or stopper needed for any flask, bottle, etc.
Insert the thin triangular gauge into the vessel and the correct stopper size is read at the top of the neck. Two scales are provided: one for rubber and vinyl stoppers and the other for corks. The back side is a circular scale to size tubing outside diameters between 4 and 52 mm.