ContactUsLinkComponent
SP Bel-Art Tubing Connector Assortment, Bel-Art Products, a part of SP
SP Bel-Art Tubing Connector Assortment, Bel-Art Products, a part of SP
Catalog # 70690-194
undefined
SP Bel-Art Tubing Connector Assortment, Bel-Art Products, a part of SP
Catalog # 70690-194
Supplier Number:  H19570-0000
Specifications
  • Description:
    22-Piece Fitting Kit
  • Cat. No.:
    70690-194
  • Supplier no.:
    195700000
Specifications
About this item
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7 mm (⅛ to ½").
  • Chemically resistant
  • Polypropylene construction
  • Autoclavable
Chemical-resistant for use with a wide range of liquids or gases. FDA-grade polypropylene for direct contact with food, drugs, and cosmetics. Autoclavable.
Ordering information: Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors.