Order Entry
United States
ContactUsLinkComponent
Mercury(II) sulfate 200 g/l in dil. sulfuric acid for COD determination, Supelco®
Toxic
Health
Corrosive
Environment
Mercury(II) sulfate 200 g/l in dil. sulfuric acid for COD determination, Supelco®
Catalog # EM1.04413.1000
Supplier:  MilliporeSigma
CAS Number:  
Toxic
Health
Corrosive
Environment
Mercury(II) sulfate 200 g/l in dil. sulfuric acid for COD determination, Supelco®
Catalog # EM1.04413.1000
Supplier:  MilliporeSigma
Supplier Number:  1.04413.1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    EM1.04413.1000
  • Supplier no.:
    1.04413.1000

Specifications

Specification Test Results

  • Concentration (HgSO₄):
    198.0 - 202.0 g/l
  • Concentration (H₂SO₄):
    173.0 - 180.0 g/l

Specification Test Results

About this item

 :
 :296.65 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
 :GHS06
 :GHS08
 :GHS05
 :GHS09
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047