



Aniline, EMSURE® for analysis, Supelco®
Catalog # EM1.01261.1000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:EM1.01261.1000
- Supplier no.:1.01261.1000
Specifications
Specification Test Results
- Solubility:36 g/l (20 °C)
- Melting Point:-6.2 °C
- Molar Mass:93.13 g/mol
- Boiling Point:184 °C (1013 hPa)
- Vapor Pressure:0.5 hPa (20 °C)
- Flash Point:76 °C
- Refractive Index:1.5863 (20 °C, 589 nm)
- Explosion Limit:1.2 - 11 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:8.8 (36 g/l, H2O, 20 °C)
- Ignition Point:540 °C
Specification Test Results
About this item
:
:93.13 g/mol
Merck Index:13,00661
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6.2 °C
Density:1.019 g/cm³ (20 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
EINECS:200-539-3
UN:1547
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:62-53-3
Formula:C₆H₅NH₂
MDL Number:MFCD00007629