Order Entry
United States
ContactUsLinkComponent
Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Catalog # EM1.00659.0100
Supplier:  MilliporeSigma
CAS Number:  
Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Catalog # EM1.00659.0100
Supplier:  MilliporeSigma
Supplier Number:  1.00659.0100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g
  • Cat. No.:
    EM1.00659.0100
  • Supplier no.:
    1.00659.0100

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    2878.17 g/mol
  • Bulk Density:
    200 - 500 kg/m3

Specification Test Results

About this item

 :
 :2896.172 g/mol
CAS Number:12027-43-9
Formula:H₄Si(W₃O₁₀)₄.xH₂O
MDL Number:MFCD00036271