SP Bel-Art Vortexer Attachments for Microcentrifuge Tubes, Bel-Art, Bel-Art Products, a part of SP
58815-252
- No. of Holes:20
- Description:Vortexer Attachment
- Environmentally Preferable:
- Cat. No.:58815-252
- Supplier no.:H37080-0020
Attachment securely holds 1.5 mL microcentrifuge tubes in an 8 or 20 capacity disk for simultaneous vortexing.
- Low-friction shaft
- Securely holds multiple microcentrifuge tubes for simultaneous mixing
Tube contents are mixed by holding the shaft against the rubber pad of a vortexer. Made of polypropylene with a 16.5 cm long Delrin® shaft and 2.54 cm dia. ball handle.