Acrodisc® Units with Mustang® Q and S Membranes, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
Disposable chromatography units with high binding capacities and fast flow rate.
- Disposable
- High binding capacity
- Fast flow rate
Mustang® Q, strong anion exchanger provides contaminant removal such as DNA viral particles, host cell proteins or endotoxins.
Mustang® S, strong cation exchanger purifies and concentrates positively-charged proteins and viral particles. Ideal for evaluation, process development and validation.
Ideal for isolation via capture and release of plasmid DNA, virus or target protein from a complex mixture. Offer protein polishing for negatively charged proteins. Purifies virus and oligo nucleotides.
Recommended flow rate: 1 to 4 ml/min.
Frequently Bought Together