Spherical Reaction Flask with Conical Flange, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Rugged spherical flask with conical neck opening of 100 mm (4")
Standard size flasks use regular Glas-Col heating mantles of equivalent capacity. Length of straight section including flange, approximately 95 mm (3 3/4"). Uses a clamp for securing flask head to flask. No Valve.
Frequently Bought Together