Power Supply, Photochemical, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Newly redesigned power supply transformers, for photochemical lamps, that supply the extra voltage and current required to initiate the arc and to reduce power for operation
Use with the Immersion Lamps by Ace Glass. The 'A' Power Supply units use with 89084-186, the 'B' with 89084-188, the 'C' with 89084-190, the 'D' with 89084-192, and the 'G' units with 89084-184.
Ordering information: Two year warranty included from date of shipment (When used under normal conditions with ACE equipment).
Frequently Bought Together