Anti-TRAP Rabbit Polyclonal Antibody
Catalog # 89418-204
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:acid phosphatase 5, tartrate resistant
- Antigen Symbol:ACP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P13686
- Antigen Synonyms:ACP5
- Storage Buffer:TRAP Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:TRAP antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:TRAP antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human TRAP. The immunogen is located within the last 50 amino acids of TRAP.
- Tested Applications:E, WB, IF
- Purification:TRAP Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89418-204
- Supplier no.:6911
Specifications
About this item
TRAP Antibody: TRAP, also known as uteroferrin, is an iron containing, glycosylated, acid phosphatase. It is the most basic of the acid phosphatases and is the only form not inhibited by L(+)-tartrate. Along with the related protein ACP2, TRAP mediates the removal of mannose 6-phosphate residues from proteins targeted to lysosomes. TRAP is present in brain at low levels, but is expressed at a much higher level in liver.
Type: Primary
Antigen: ACP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse