Anti-OCC1 Rabbit Polyclonal Antibody
89416-872
: Prosci
- Antibody Type:Primary
- Antigen Name:chromosome 12 open reading frame 75
- Antigen Symbol:C12orf75
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q8TAD7
- Antigen Synonyms:C12orf75
- Storage Buffer:OCC-1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:OCC-1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:OCC-1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human OCC-1. The immunogen is located within amino acids 10 - 60 of OCC-1.
- Tested Applications:E, WB, IHC, IF
- Purification:OCC-1 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89416-872
- Supplier no.:4997
OCC-1 Antibody: OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
Type: Primary
Antigen: C12orf75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human