- Antibody Type:Primary
- Antigen Name:toll-like receptor 7
- Antigen Symbol:Tlr7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P58681
- Antigen Synonyms:Tlr7
- Storage Buffer:TLR7 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 116 kDa Observed: 135 kDa
- Storage Temperature:TLR7 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:TLR7 antibody was raised against a peptide corresponding to 14 amino acids near the center of mouse TLR7. The immunogen is located within amino acids 420 - 470 of TLR7.
- Tested Applications:Immunocytochemistry
- Purification:TLR7 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-658
- Supplier no.:3269
TLR7 Antibody: Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. These proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors. TLR7, like TLRs 3, 8, and 9, is localized in intracellular acidic compartments such as the phagolysosome and will recognize some single-stranded RNA viruses such as vesicular stomatitis virus (VSV) and influenza virus. Activation of TLR7 by VSV results in stimulation of the immune response including IFNalpha secretion, suggesting the importance of TLR7 in virus recognition.
Type: Primary
Antigen: Tlr7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse