SP Bel-Art Spinbar® Positioner/Retriever, Bel-Art Products, a part of SP
58948-285
- Ø:15.9 mm (5/8 ")
- Length:305 mm (12")
- Cat. No.:58948-285
- Supplier no.:H377800000
Position a Spinbar® Magnetic Stirring Bar exactly where desired, without splashing, using the double duty Spinbar® Positioner/Retriever.
- Polyethylene construction
- Easy to use
- Can position stirring bars without splashing
Users can also remove bars without touching or pouring out the contents of the vessel. To pick up a stirring bar, hold the polyethylene retriever down; to release a stirring bar, slide the retriever up inside the tube.