Nunc® Low Profile 5.0 ml Universal Tubes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific Nunc™ low profile 5.0 ml externally-threaded universal tubes provide a robust solution for solid or liquid sample storage.
- Tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures
Available in configurations that are designed to meet most tracking and packaging requirements, the low profile 5.0 ml universal tube features 2D barcodes, graduations, a white patch for labeling, recommended working volume of 4.5 ml, and a wide mouth design for easy addition and extraction of sample material.
The externally-threaded cap design minimizes contamination risk in manual handling procedures.
Certifications: IATA. SAL 10⁻⁶.
Related Information
Certificates
Search by Lot NumberAR-14TF 5.0 UniversalTubes VWR 0828
Datasheet
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Frequently Bought Together