SP Bel-Art Spinfin® Magnetic Stirring Bars, Bel-Art Products, a part of SP
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
- Maintains its position in a vessel and will not wobble
- PTFE low-friction coating
- Durable and inert
- Autoclave safe
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Frequently Bought Together