SP Bel-Art Height Adjustable Water Bath Rack, Bel-Art Products, a part of SP
Catalog # 89042-246
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Height Adjustable Water Bath Rack
- Cat. No.:89042-246
- Supplier no.:F188820000
Specifications
About this item
Height-Adjustable Racks securely hold up to six tubes or vials at any desired depth in a water bath or beaker.
- Flexible grip
- Adjustable height prevents submersion problems
- Compact footprint
- Stable base
- Autoclavable
The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1.5 mL to 15 mL.
Unlike floating racks, this rack’s adjustable height prevents submersion and sinking problems that might occur with heavier or unbalanced tubes. The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing. All parts are steam autoclavable at 121 °C (250 °F).