About this item
The Wizard MagneSil Plasmid DNA Purification System provides a simple, reliable method for rapid isolation of plasmid DNA in a 96-well, high-throughput format.
- Rapid Isolation of Plasmid DNA in 96-Well Format
- Process multiple plates without user intervention
- Validated automated methods available
The Wizard MagneSil Plasmid DNA Purification System provides a simple and reliable method for the rapid isolation of plasmid DNA in a 96-well, high-throughput format. The purified plasmid can be used directly for automated fluorescent sequencing, such as with BigDye terminator sequencing chemistry, as well as for other standard molecular biology techniques including restriction enzyme digestion. The use of the MagneSil Paramagnetic Particles for lysate clearing (BLUE) as well as DNA capture (RED) circumvents the need for centrifugation or vacuum manifolds, making the system ideal for full automation on a Beckman Coulter or Tecan instrument.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: