BI-OK® EO Gas Biological Indicator Vials, Propper Manufacturing
Catalog # 58752-812
Supplier: Propper
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:BI-O.K.® EO-Gas Vials
- Cat. No.:58752-812
- Supplier no.:269301
Specifications
About this item
BI-OK® biological indicators are a reliable method to test ethylene oxide sterilization cycles.
- Self-Contained biological indicator vials for ethylene oxide
- Vials provide test results in 48 hours
Vials are inoculated with Geobacillus stearothermophilus spores to challenge steam sterilization. Use in steam test pack during cycle, after sterilization cycle activate media by crushing the glass vial with the vial activator and incubate at 57 °C. A color change in the media indicates spore growth and cycle failure.