Once-A-Day® Bowie Dick Test Pak, Propper Manufacturing
Catalog # 15425-014
Supplier: Propper
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Once-A-Day® Bowie Dick Test Pak
- Environmentally Preferable:
- Cat. No.:15425-014
- Supplier no.:266103
Specifications
About this item
Once-A-Day Test Pak® is a pre-assembled disposable Bowie and Dick test for daily monitoring of pre-vacuum sterilizers at 270 to 276 °F.
- Pre-assembled disposable Bowie-Dick test pack
- Distinct white to black color change
- External process indicator helps distinguish exposed pack
Bowie-Dick tests provide a challenge for pre-vacuum sterilizers at the start of each day, or after routine or other maintenance. Once-A-Day® detects low temperature/pressure conditions, wet and superheated steam and other technical malfunctions. For use in pre-vacuum steam sterilizers at 270 to 276 °F.
Certifications: AAMI / ISO 11140-1:2010, Class 2 AAMI / ISO 11140-5:2007