Order Entry
United States
ContactUsLinkComponent
 
SP Bel-Art Poxygrid® Tube Slant Rack, 50 ml, Bel-Art Products, a part of SP
  89009-200
undefined
SP Bel-Art Poxygrid® Tube Slant Rack, 50 ml, Bel-Art Products, a part of SP
  89009-200
 :  F18794-0028

 

  • No. of Holes:
    8
  • Description:
    Poxygrid® 50mL Tube Slant Rack
  • Cat. No.:
    89009-200
  • L×W×H:
    15.2 x 12.1 x 12.1 cm (6 x 4³/₄ x 4³/₄")
  • Supplier no.:
    187940028

 

 

Angled rack is useful for keeping tube caps and marked contents easily readable, frequently used reagents at an ergonomic angle, or draining excess supernatant fluid from pellets.

  • Angled rack design
  • Epoxy-coated steel wire construction
  • Mountable using built-in mounting points
  • Unaffected by organic solvents and weak acids and alkalis
  • Autoclavable

Each rack holds eight 50 mL conical tubes or other vessels up to 30 mm in diameter.

To keep samples off the bench but still immediately available, the rack can be attached to the underside of a shelf using built-in mounting points (hardware not included). The steel wire is epoxy coated for long life and easy cleaning. It is unaffected by organic solvents, salts, weak acids, and weak alkalis, and can be steam autoclaved at 121°C (250°F).