Uri-Pak™ Urine Collection Kit, Globe Scientific
Catalog # 89497-592
Supplier: Globe Scientific
Specifications
- Description:Uri-Pak™ Urine Collection Kit
- Cat. No.:89497-592
- Supplier no.:112019R
Specifications
About this item
The Uri-Pak™ Urine Collection Kit offers all the components necessary for specimen collection, handling, transport, and testing
The Kit consists of 12mL flared top, polystyrene urine tube, yellow polyethylene snap caps with sanitary grip, collection cups (3.5oz.), adhesive ID labels, and a rack.
Uri-Pak™ is used in hospitals, laboratories, physicians' offices and nursing homes throughout the world.