Specifications
- Interface:USB
- Description:Hanna® multiparameter environmental testing meter with GPS
- Cat. No.:470213-274
- Supplier no.:HI9829-10041
Specifications
About this item
This device is virtually a complete environmental testing outfit. Measure up to 13 different water quality parameters, including pH, pH/mV, ORP, TDS, mg/L dissolved oxygen, EC, absolute EC, percent saturation, salinity, seawater specific gravity, atmospheric pressure, and temperature. In addition to the readings, the meter also records the exact location where measurements were recorded using a built-in 12 channel GPS receiver. Accuracy is guaranteed within a 30 feet range, and detailed coordinate information can be viewed immediately on the display and then later transferred to a computer using the included software.
Hanna’s exclusive Fast Tracker™ Tag Identification System, included, simplifies test logging too. When the matching connector on the meter contacts the location button, measurements are logged and labeled with the alphnumeric user-entered location ID. The location, date, time, and measurements are then logged into the meter, which can later be transferred to a PC. A graphic, backlit display automatically sizes digits to fit the screen and allows full configuration of each parameter along with on-screen graphing capability. Additionally, each parameter is supported by on-screen help, both in calibration and measurement modes. The rugged, impact-resistant unit is also waterproofed to withstand 30 minutes of immersion under 1 m of water without damage.
This versatile, comprehensive meter comes complete with quick calibration solution, a probe maintenance kit, rechargeable C batteries, power adapter and cable, USB cable, software, and instructions — all packaged in a rugged carrying case.
Specifications:
pH range: 0 to 14 pH, resolution: 0.01 pH, accuracy: ±0.02 pH. mV range: ±600.0 mV, resolution: 0.1 mV, accuracy: ±0.5 mV. ORP range: ±2000.0 mV, resolution: 0.1 mV, accuracy: ±1.0 mV. DO range: 0 to 500%/0 to 50.0 mg/L, resolution: 0.1%/0.01 mg/L, accuracy: ±1.5% of reading 0 to 300%, ±3% of reading 300 to 500%. Conductivity range: 0 to 200.000 mS/cm (absolute EC up to 400 mS/cm), resolution: variable, 1 μS/cm from 0 to 9999 μS/cm, accuracy: ±1% of reading. TDS range: 0 to 400,000 mg/L or ppm, resolution: variable, 1 mg/L (ppm), accuracy: ±1% of reading. Temperature range: −5 to 55 °C/23 to 131 °F, resolution: 1 °C/F, accuracy ±0.15 °C or ±0.27 °F. Temperature compensation: −5 to +55 °C. Meter size: 8 3/4" × 4 1/2" × 2 1/4"; diameter: 6 3/5" × 1 4/5". Probe size: 10 2/3" × 1 3/4" diameter; weight: 26.5 oz. NEW and Improved model features: Updated electronics and more stable and durable cable connection; new menu and navigation features; custom-fit case; and new improved flow cell.