Order Entry
United States
ContactUsLinkComponent
Slide Dispenser for 72 Slides, Globe Scientific
Slide Dispenser for 72 Slides, Globe Scientific
Catalog # 89510-754
Supplier:  Globe Scientific
undefined
Slide Dispenser for 72 Slides, Globe Scientific
Catalog # 89510-754
Supplier:  Globe Scientific
Supplier Number:  513000

Specifications

  • Description:
    Slide Dispenser for 72 Slides
  • Cat. No.:
    89510-754
  • Supplier no.:
    513000

Specifications

About this item

The Globe microscope slide dispenser is designed for use with 25 x 75 mm and 3 x 1" microscope slides

To use, simply insert one full box of 72 slides into the holder. Turn the dial to dispense one slide at a time. This convenient dispenser prevents slides from getting dirty, dusty and scratched.