Specifications
- Size:250 Units
- Cat. No.:80053-704
- Supplier no.:525200-250U
Specifications
About this item
Native phospholipase D from Streptomyces chromofuscus
Catalyzes the hydrolysis of lecithin to phosphatidic acid and choline. Coupled with choline oxidase, it can also be used for the determination of phospholipids in serum.