ContactUsLinkComponent
Hardened Steel Mortar, Retsch®
Hardened Steel Mortar, Retsch®
Catalog # 14213-042
Supplier:  Retsch
Hardened Steel Mortar, Retsch®
Catalog # 14213-042
Supplier:  Retsch
Supplier Number:  024600018
Specifications
  • Description:
    Hardened Steel Mortar
  • Cat. No.:
    14213-042
  • Supplier no.:
    024600018
Specifications
About this item
Designed for use with Retsch® RM200 Mortar Grinder, this hardened steel mortar is intended for use in less demanding applications, or under rough conditions
Material composition is approximately 86% Fe, 11.5% Cr, and trace amounts of C, Mn, Si, P, and S. Density is approximately 7700kg/m3.