Order Entry
United States
ContactUsLinkComponent
Hydranal® Composite reagents for Volumetric Karl Fischer Titration, Aqua Solutions
Hydranal® Composite reagents for Volumetric Karl Fischer Titration, Aqua Solutions
Catalog # 101444-676
Supplier:  Aqua Solutions
CAS Number:  
Hydranal® Composite reagents for Volumetric Karl Fischer Titration, Aqua Solutions
Catalog # 101444-676
Supplier:  Aqua Solutions
Supplier Number:  ALD-34816-1
CAS Number:  

Specifications

  • Description:
    Hydranal® Composite 5K
  • Size:
    1 L
  • Cat. No.:
    101444-676
  • Supplier no.:
    ALD-34816-1

Specifications

About this item

Component reagent for volumetric titration.