Human NCLN Peptide (18 aa near N-terminus)
89421-278
: Prosci
:
- Species:Human
- Size:0.05 mg
- Storage Conditions:Store at –20 °C, stable for one year.
- Amino Acid Number:18 aa near N-terminus
- Protein/Peptide Name:NCLN Peptide (18 aa near N-terminus)
- Tested Applications:Blocking
- Cat. No.:89421-278
- Supplier no.:6789P
NCLN peptide is used for blocking the activity of NCLN antibody.